Product Details

SNP ID
rs8035615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64912739 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAATCCCGGGGCTTTCCCTCAAA[A/T]GCGCATAGTGCAGCTCACTGGATTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKDD1A PubMed Links

Gene Details

Gene
ANKDD1A
Gene Name
ankyrin repeat and death domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182703.5 Intron NP_874362.3

View Full Product Details