Product Details

SNP ID
rs1727314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123156117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGGCAGAGAGCTACTCTGAGAG[A/C]CCTCCAGAGAGGGCTCTCCAATTGT
Phenotype
MIM: 605501
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MPHOSPH9 PubMed Links

Gene Details

Gene
MPHOSPH9
Gene Name
M-phase phosphoprotein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022782.3 4243 Intron NP_073619.3
XM_011537740.2 4243 Intron XP_011536042.1
XM_011537741.2 4243 Intron XP_011536043.1
XM_017018673.1 4243 Intron XP_016874162.1
XM_017018674.1 4243 Intron XP_016874163.1
XM_017018675.1 4243 Intron XP_016874164.1
XM_017018676.1 4243 Intron XP_016874165.1
XM_017018677.1 4243 UTR 3 XP_016874166.1
XM_017018678.1 4243 Intron XP_016874167.1
XM_017018679.1 4243 Intron XP_016874168.1
XM_017018680.1 4243 Intron XP_016874169.1
XM_017018681.1 4243 Intron XP_016874170.1

View Full Product Details