Product Details

SNP ID
rs9858949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183645039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTTTTTGAACCTGAAATGCAA[C/T]AGGAATTTAAACTAGATTAAGCTGG
Phenotype
MIM: 611295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL24 PubMed Links

Gene Details

Gene
KLHL24
Gene Name
kelch like family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017644.3 1179 Intron NP_060114.2
XM_005247552.2 1179 Intron XP_005247609.1
XM_005247553.1 1179 Intron XP_005247610.1
XM_005247554.2 1179 Intron XP_005247611.1
XM_005247555.1 1179 Intron XP_005247612.1
XM_005247556.1 1179 Intron XP_005247613.1
XM_011512938.2 1179 Intron XP_011511240.1
XM_017006650.1 1179 Intron XP_016862139.1
XM_017006651.1 1179 Intron XP_016862140.1
XM_017006652.1 1179 Intron XP_016862141.1
XM_017006653.1 1179 Intron XP_016862142.1
XM_017006654.1 1179 Intron XP_016862143.1
XM_017006655.1 1179 Intron XP_016862144.1
XM_017006656.1 1179 Intron XP_016862145.1
XM_017006657.1 1179 Intron XP_016862146.1
XM_017006658.1 1179 Intron XP_016862147.1
XM_017006659.1 1179 UTR 5 XP_016862148.1
XM_017006660.1 1179 Intron XP_016862149.1
XM_017006661.1 1179 Intron XP_016862150.1
XM_017006662.1 1179 Intron XP_016862151.1
XM_017006663.1 1179 Intron XP_016862152.1

View Full Product Details