Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017644.3 | 1890 | Intron | NP_060114.2 | ||
XM_005247552.2 | 1890 | Intron | XP_005247609.1 | ||
XM_005247553.1 | 1890 | Intron | XP_005247610.1 | ||
XM_005247554.2 | 1890 | Intron | XP_005247611.1 | ||
XM_005247555.1 | 1890 | Intron | XP_005247612.1 | ||
XM_005247556.1 | 1890 | Intron | XP_005247613.1 | ||
XM_011512938.2 | 1890 | Intron | XP_011511240.1 | ||
XM_017006650.1 | 1890 | Intron | XP_016862139.1 | ||
XM_017006651.1 | 1890 | Intron | XP_016862140.1 | ||
XM_017006652.1 | 1890 | Intron | XP_016862141.1 | ||
XM_017006653.1 | 1890 | Intron | XP_016862142.1 | ||
XM_017006654.1 | 1890 | Intron | XP_016862143.1 | ||
XM_017006655.1 | 1890 | Intron | XP_016862144.1 | ||
XM_017006656.1 | 1890 | Intron | XP_016862145.1 | ||
XM_017006657.1 | 1890 | Intron | XP_016862146.1 | ||
XM_017006658.1 | 1890 | Intron | XP_016862147.1 | ||
XM_017006659.1 | 1890 | UTR 5 | XP_016862148.1 | ||
XM_017006660.1 | 1890 | Intron | XP_016862149.1 | ||
XM_017006661.1 | 1890 | Intron | XP_016862150.1 | ||
XM_017006662.1 | 1890 | Intron | XP_016862151.1 | ||
XM_017006663.1 | 1890 | Intron | XP_016862152.1 |