Product Details

SNP ID
rs3093047
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:109915150 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
ACTACTTCTCCTGTATGTAGTTGCT[A/G]TAGCAATCCAAGGTAGACCAAGAGC
Phenotype
MIM: 120420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF1 PubMed Links

Gene Details

Gene
CSF1
Gene Name
colony stimulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000757.5 Intron NP_000748.3
NM_172210.2 Intron NP_757349.1
NM_172211.3 Intron NP_757350.1
NM_172212.2 Intron NP_757351.1
XM_017000369.1 Intron XP_016855858.1

View Full Product Details