Product Details

SNP ID
rs34204663
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:131010500 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTGTAATGGTTTTCCATCCTAGACC[A/G]GAACTGTTGGGGAGGTGACACACCA
Phenotype
MIM: 604349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMC3 PubMed Links

Gene Details

Gene
LAMC3
Gene Name
laminin subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006059.3 Intron NP_006050.3
XM_006716921.2 Intron XP_006716984.1
XM_011518121.1 Intron XP_011516423.1

View Full Product Details