Product Details

SNP ID
rs2811789
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136799504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCCGGCCAGCTAGGGGACACCC[A/G]GGCCCTCCTCTGCATACCCCCCTAC
Phenotype
MIM: 615955 MIM: 610615
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CCDC183 PubMed Links
Additional Information
For this assay, SNP(s) [rs115824302] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CCDC183
Gene Name
coiled-coil domain containing 183
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039374.4 Intron NP_001034463.4
Gene
CCDC183-AS1
Gene Name
CCDC183 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RABL6
Gene Name
RAB, member RAS oncogene family-like 6
There are no transcripts associated with this gene.

Gene
TMEM141
Gene Name
transmembrane protein 141
There are no transcripts associated with this gene.

View Full Product Details