Product Details

SNP ID
rs2017583
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:16023533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCCCAGCCCCCAGTGTGAGGCA[A/G]GCCAGCAGAGGGTAGCTCCAGGCTC
Phenotype
MIM: 602024 MIM: 610692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKA PubMed Links

Gene Details

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042704.1 Intron NP_001036169.1
NM_001257139.1 Intron NP_001244068.1
NM_004070.3 Intron NP_004061.3
Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
There are no transcripts associated with this gene.

View Full Product Details