Product Details
- SNP ID
-
rs1131978
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:93612977 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAGCAGGGGGTTCACAGGGCCCT[A/G]CAGAGGCTGGCCCACCTGCTCCAGG
- Phenotype
-
MIM: 600827
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6C
PubMed Links
Gene Details
- Gene
- PDE6C
- Gene Name
- phosphodiesterase 6C
View Full Product Details