Product Details

SNP ID
rs636041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77875363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTTGCAAATTGGCACAAATCT[A/G]CCTGGAGACATGTTTTGTTTGTCTC
Phenotype
MIM: 611348 MIM: 608522
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AAMDC PubMed Links

Gene Details

Gene
AAMDC
Gene Name
adipogenesis associated Mth938 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316957.1 4442 Intron NP_001303886.1
NM_001316958.1 4442 Intron NP_001303887.1
NM_001316960.1 4442 Intron NP_001303889.1
NM_001316961.1 4442 Intron NP_001303890.1
NM_001316962.1 4442 Intron NP_001303891.1
NM_024684.3 4442 Intron NP_078960.1
XM_011544968.2 4442 Intron XP_011543270.1
XM_011544969.2 4442 Intron XP_011543271.1
XM_017017620.1 4442 Intron XP_016873109.1
Gene
INTS4
Gene Name
integrator complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033547.3 4442 Intron NP_291025.3
XM_011545352.2 4442 Intron XP_011543654.1
XM_011545353.2 4442 Intron XP_011543655.1
XM_017018560.1 4442 UTR 3 XP_016874049.1
Gene
RSF1
Gene Name
remodeling and spacing factor 1
There are no transcripts associated with this gene.

View Full Product Details