Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022782.3 | 4074 | Intron | NP_073619.3 | ||
XM_011537740.2 | 4074 | UTR 3 | XP_011536042.1 | ||
XM_011537741.2 | 4074 | Intron | XP_011536043.1 | ||
XM_017018673.1 | 4074 | Intron | XP_016874162.1 | ||
XM_017018674.1 | 4074 | Intron | XP_016874163.1 | ||
XM_017018675.1 | 4074 | Intron | XP_016874164.1 | ||
XM_017018676.1 | 4074 | Intron | XP_016874165.1 | ||
XM_017018677.1 | 4074 | Intron | XP_016874166.1 | ||
XM_017018678.1 | 4074 | Intron | XP_016874167.1 | ||
XM_017018679.1 | 4074 | Intron | XP_016874168.1 | ||
XM_017018680.1 | 4074 | Intron | XP_016874169.1 | ||
XM_017018681.1 | 4074 | Intron | XP_016874170.1 |