Product Details

SNP ID
rs2294972
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:37614028 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTCATCTGTCTGTCTACACCCACA[A/C]TCAGGTCTTGAGGCTGTCTGGGCAG
Phenotype
MIM: 606004
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GGA1 PubMed Links

Gene Details

Gene
GGA1
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001560.2 Intron NP_001001560.1
NM_001172687.1 Intron NP_001166158.1
NM_001172688.1 Intron NP_001166159.1
NM_013365.4 Intron NP_037497.1
XM_005261517.3 Intron XP_005261574.1
XM_005261518.1 Intron XP_005261575.1
XM_005261520.1 Intron XP_005261577.1
XM_005261522.1 Intron XP_005261579.1
XM_006724229.1 Intron XP_006724292.1
XM_011530122.1 Intron XP_011528424.1
XM_011530123.1 Intron XP_011528425.1
XM_011530124.1 Intron XP_011528426.1
XM_017028760.1 Intron XP_016884249.1
XM_017028761.1 Intron XP_016884250.1
XM_017028762.1 Intron XP_016884251.1
XM_017028763.1 Intron XP_016884252.1
XM_017028764.1 Intron XP_016884253.1
XM_017028765.1 Intron XP_016884254.1

View Full Product Details