Product Details

SNP ID
rs743578
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:2430234 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCCCTCCCCTGCCCTGTTGGCC[C/T]TGCACAGTCAGGAACTGAACTGCGG
Phenotype
MIM: 150341 MIM: 607383 MIM: 610477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMNB2 PubMed Links

Gene Details

Gene
LMNB2
Gene Name
lamin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032737.3 2575 UTR 3 NP_116126.3
Gene
MIR7108
Gene Name
microRNA 7108
There are no transcripts associated with this gene.

Gene
TIMM13
Gene Name
translocase of inner mitochondrial membrane 13
There are no transcripts associated with this gene.

Gene
TMPRSS9
Gene Name
transmembrane protease, serine 9
There are no transcripts associated with this gene.

View Full Product Details