Product Details

SNP ID
rs34195752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8220139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCGGCTCTCCTCGCCCCCTCC[C/T]ACCCCTCGGCTGTGTGTCCCCTTAT
Phenotype
MIM: 615334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CERS4 PubMed Links

Gene Details

Gene
CERS4
Gene Name
ceramide synthase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024552.2 Intron NP_078828.2
XM_011528290.2 Intron XP_011526592.1
XM_011528291.2 Intron XP_011526593.1
XM_011528292.2 Intron XP_011526594.1
XM_011528293.2 Intron XP_011526595.1
XM_011528294.2 Intron XP_011526596.1
XM_011528295.2 Intron XP_011526597.1
XM_017027303.1 Intron XP_016882792.1
XM_017027304.1 Intron XP_016882793.1
XM_017027305.1 Intron XP_016882794.1

View Full Product Details