Product Details

SNP ID
rs12436310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96044988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACCACAACAGGTTCTGAAGACCC[A/G]TAAAGGATAAGCATCTTGGCCCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf132 PubMed Links

Gene Details

Gene
C14orf132
Gene Name
chromosome 14 open reading frame 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252507.2 Intron NP_001239436.1
NM_001282463.1 Intron NP_001269392.1
NM_001282464.1 Intron NP_001269393.1
NM_001289139.1 Intron NP_001276068.1
XM_017021469.1 Intron XP_016876958.1

View Full Product Details