Product Details

SNP ID
rs942251
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:46490605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACGCGAACCCCGCAGTGCTCC[C/G]GGCATTGCCCGGGTCTGGCTTTGGT
Phenotype
MIM: 607410
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DMBX1 PubMed Links

Gene Details

Gene
DMBX1
Gene Name
diencephalon/mesencephalon homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147192.2 71 Intron NP_671725.1
NM_172225.1 71 Intron NP_757379.1
XM_011540668.2 71 UTR 5 XP_011538970.1
XM_017000289.1 71 Intron XP_016855778.1

View Full Product Details