Product Details

SNP ID
rs708243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8366312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAGTGATTTGTATATAACTGG[C/T]GGCAGGGGGAGGGGGGCGGTTGGAG
Phenotype
MIM: 603402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOX3 PubMed Links

Gene Details

Gene
ACOX3
Gene Name
acyl-CoA oxidase 3, pristanoyl
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101667.1 2776 UTR 3 NP_001095137.1
NM_003501.2 2776 UTR 3 NP_003492.2
XM_005248011.4 2776 UTR 3 XP_005248068.1
XM_005248013.3 2776 UTR 3 XP_005248070.1
XM_011513565.2 2776 UTR 3 XP_011511867.1
XM_011513566.1 2776 UTR 3 XP_011511868.1

View Full Product Details