Product Details

SNP ID
rs11592969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96595055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACTAATCCCTCTAGTCTTTTTC[A/G]GTTGGTTGTGTGTGACCAGCCCTAG
Phenotype
MIM: 607942 MIM: 616872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3AP1 PubMed Links

Gene Details

Gene
PIK3AP1
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152309.2 2615 UTR 3 NP_689522.2
XM_005269498.1 2615 UTR 3 XP_005269555.1
XM_005269499.1 2615 UTR 3 XP_005269556.1
XM_011539248.1 2615 UTR 3 XP_011537550.1
XM_011539249.1 2615 UTR 3 XP_011537551.1
Gene
TM9SF3
Gene Name
transmembrane 9 superfamily member 3
There are no transcripts associated with this gene.

View Full Product Details