Product Details

SNP ID
rs9378720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2624140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCCCTGCAGAAAAGGGAGACAGA[A/G]GCAGAGCGTGGGTCAGAGCCACACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01600 PubMed Links

Gene Details

Gene
LINC01600
Gene Name
long intergenic non-protein coding RNA 1600
There are no transcripts associated with this gene.

View Full Product Details