Product Details

SNP ID
rs11754153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33624434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCAGCAGCTTCAGCGTCACCA[A/G]GAAGTTTGTCAGAAATGCAGAATCT
Phenotype
MIM: 147267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPR3 PubMed Links

Gene Details

Gene
ITPR3
Gene Name
inositol 1,4,5-trisphosphate receptor type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002224.3 Intron NP_002215.2
XM_011514577.2 Intron XP_011512879.1
XM_017010832.1 Intron XP_016866321.1
Gene
LOC101929188
Gene Name
uncharacterized LOC101929188
There are no transcripts associated with this gene.

View Full Product Details