Product Details

SNP ID
rs16822379
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:184058542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGTAGTAGGCATACTTTTATCTA[C/T]TTTTACTTTTAGTGTATTTTATAAT
Phenotype
MIM: 608756
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COLGALT2 PubMed Links

Gene Details

Gene
COLGALT2
Gene Name
collagen beta(1-O)galactosyltransferase 2
There are no transcripts associated with this gene.

Gene
TSEN15
Gene Name
tRNA splicing endonuclease subunit 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127394.3 Intron NP_001120866.1
NM_001300764.1 Intron NP_001287693.1
NM_001300766.1 Intron NP_001287695.1
NM_052965.3 Intron NP_443197.1
XM_006711148.1 Intron XP_006711211.1
XM_011509139.2 Intron XP_011507441.1
XM_017000228.1 Intron XP_016855717.1
XM_017000229.1 Intron XP_016855718.1
XM_017000230.1 Intron XP_016855719.1

View Full Product Details