Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130912.2 | 165 | Intron | NP_001124384.1 | ||
NM_001320831.1 | 165 | Intron | NP_001307760.1 | ||
NM_006704.4 | 165 | Intron | NP_006695.1 | ||
XM_011534891.2 | 165 | UTR 5 | XP_011533193.1 | ||
XM_011534893.2 | 165 | UTR 5 | XP_011533195.1 | ||
XM_017020365.1 | 165 | Intron | XP_016875854.1 | ||
XM_017020366.1 | 165 | Intron | XP_016875855.1 | ||
XM_017020367.1 | 165 | UTR 5 | XP_016875856.1 | ||
XM_017020368.1 | 165 | UTR 5 | XP_016875857.1 |