Product Details

SNP ID
rs7210713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42402983 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGGAGCAACACTCCCTTCACCC[C/G]CAAAGATTCAGGAAAAGCACCCCAA
Phenotype
MIM: 603198
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTRF PubMed Links

Gene Details

Gene
PTRF
Gene Name
polymerase I and transcript release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012232.5 3100 UTR 3 NP_036364.2
XM_005257242.3 3100 Intron XP_005257299.1

View Full Product Details