Product Details

SNP ID
rs12360577
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:28108928 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATCGGAAATACGTTTGCTAAAAT[A/C]ACTTTATTTGGCAGGGTGTTTTTAA
Phenotype
MIM: 611271
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF18A PubMed Links

Gene Details

Gene
KIF18A
Gene Name
kinesin family member 18A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031217.3 Intron NP_112494.3
XM_017018379.1 Intron XP_016873868.1
XM_017018380.1 Intron XP_016873869.1
Gene
METTL15
Gene Name
methyltransferase like 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113528.1 Intron NP_001107000.1
NM_001297775.1 Intron NP_001284704.1
NM_152636.2 Intron NP_689849.2
XM_011519935.2 Intron XP_011518237.1
XM_011519937.2 Intron XP_011518239.1
XM_011519940.2 Intron XP_011518242.1
XM_011519941.2 Intron XP_011518243.1
XM_017017297.1 Intron XP_016872786.1
XM_017017298.1 Intron XP_016872787.1
XM_017017299.1 Intron XP_016872788.1
XM_017017300.1 Intron XP_016872789.1
XM_017017301.1 Intron XP_016872790.1
XM_017017302.1 Intron XP_016872791.1
XM_017017303.1 Intron XP_016872792.1

View Full Product Details