Product Details

SNP ID
rs980099
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:107068757 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGTCTGTCATAGAAACTGAAATT[C/T]GCATATTATTCTTCCAGTTGGATTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM41 PubMed Links

Gene Details

Gene
RBM41
Gene Name
RNA binding motif protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171080.1 Intron NP_001164551.1
NM_001324242.1 Intron NP_001311171.1
NM_001324243.1 Intron NP_001311172.1
NM_001324244.1 Intron NP_001311173.1
NM_018301.4 Intron NP_060771.3
XM_011530980.2 Intron XP_011529282.1
XM_011530981.2 Intron XP_011529283.1

View Full Product Details