Product Details

SNP ID
rs1887197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93721486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACGTCTACTCCTTTGTTGCTCTG[C/T]GAAGCACTCATGGGATACTCAGCAA
Phenotype
MIM: 613851
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRIMA1 PubMed Links

Gene Details

Gene
PRIMA1
Gene Name
proline rich membrane anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178013.3 462 Silent Mutation TCA,TCG S140S NP_821092.1
XM_011536456.2 462 Silent Mutation TCA,TCG S140S XP_011534758.1

View Full Product Details