Product Details
- SNP ID
-
rs1256947
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:201406685 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCTCTCCAGAGGCCAGGCCCCC[A/G]GCCCAGGCCCCATTCTTGGCTACAA
- Phenotype
-
MIM: 602314
MIM: 191042
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116649880] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAD1
- Gene Name
- ladinin 1
There are no transcripts associated with this gene.
- Gene
- TNNI1
- Gene Name
- troponin I1, slow skeletal type
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003281.3 |
3225 |
UTR 3 |
|
|
NP_003272.3 |
View Full Product Details