Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142685.1 | 8486 | UTR 3 | NP_001136157.1 | ||
NM_014715.3 | 8486 | UTR 3 | NP_055530.2 | ||
XM_005271736.4 | 8486 | UTR 3 | XP_005271793.2 | ||
XM_011543072.2 | 8486 | UTR 3 | XP_011541374.1 | ||
XM_011543073.2 | 8486 | UTR 3 | XP_011541375.2 | ||
XM_011543075.2 | 8486 | UTR 3 | XP_011541377.1 | ||
XM_017018595.1 | 8486 | UTR 3 | XP_016874084.1 | ||
XM_017018596.1 | 8486 | UTR 3 | XP_016874085.1 | ||
XM_017018597.1 | 8486 | Intron | XP_016874086.1 |