Product Details

SNP ID
rs11958638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:129102240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAGAATGTCTATGAAAGGTAGG[A/G]TATTTTTTATTCAGTAAATTGTAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISOC1 PubMed Links

Gene Details

Gene
ISOC1
Gene Name
isochorismatase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016048.2 Intron NP_057132.2
Gene
MIR4633
Gene Name
microRNA 4633
There are no transcripts associated with this gene.

View Full Product Details