Product Details

SNP ID
rs2334293
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:47747063 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAGGCTTTTGTGGTGTGGAATGGC[C/G]GTTATTCATTGTTAGTCAGCTTACG
Phenotype
MIM: 605890 MIM: 605691
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EHD2 PubMed Links

Gene Details

Gene
EHD2
Gene Name
EH domain containing 2
There are no transcripts associated with this gene.

Gene
GLTSCR2
Gene Name
glioma tumor suppressor candidate region gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015710.4 Intron NP_056525.2
Gene
GLTSCR2-AS1
Gene Name
GLTSCR2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SNORD23
Gene Name
small nucleolar RNA, C/D box 23
There are no transcripts associated with this gene.

View Full Product Details