Product Details

SNP ID
rs2047563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:140680357 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTCCTGCCTCACTCTGGTCTCA[C/T]TCATCCCTCTCCATCTTAATCTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM42 PubMed Links

Gene Details

Gene
TRIM42
Gene Name
tripartite motif containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152616.4 Intron NP_689829.3
XM_011512740.2 Intron XP_011511042.1

View Full Product Details