Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001123.3 | Intron | NP_001114.2 | |||
NM_001202449.1 | Intron | NP_001189378.1 | |||
NM_001202450.1 | Intron | NP_001189379.1 | |||
NM_006721.3 | Intron | NP_006712.2 | |||
XM_017015698.1 | Intron | XP_016871187.1 | |||
XM_017015699.1 | Intron | XP_016871188.1 | |||
XM_017015700.1 | Intron | XP_016871189.1 | |||
XM_017015701.1 | Intron | XP_016871190.1 | |||
XM_017015702.1 | Intron | XP_016871191.1 | |||
XM_017015703.1 | Intron | XP_016871192.1 | |||
XM_017015704.1 | Intron | XP_016871193.1 | |||
XM_017015705.1 | Intron | XP_016871194.1 | |||
XM_017015706.1 | Intron | XP_016871195.1 |