Product Details

SNP ID
rs6874059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79384358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACTACAGAAATACAAAAGATTAC[A/G]AGAGACTATTATGAGGTATACATGA
Phenotype
MIM: 604798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOMER1 PubMed Links

Gene Details

Gene
HOMER1
Gene Name
homer scaffolding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277077.1 Intron NP_001264006.1
NM_001277078.1 Intron NP_001264007.1
NM_004272.4 Intron NP_004263.1
XM_017010059.1 Intron XP_016865548.1

View Full Product Details