Product Details

SNP ID
rs3912000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202878969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTGATGAAAGGTTTTGTCTAAA[A/G]AATTTATTCTAATCTACACATGAGG
Phenotype
MIM: 616620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICA1L PubMed Links

Gene Details

Gene
ICA1L
Gene Name
islet cell autoantigen 1 like
There are no transcripts associated with this gene.

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 3790 Intron NP_060726.3
XM_011511469.2 3790 UTR 3 XP_011509771.1

View Full Product Details