Product Details
- SNP ID
-
rs36033494
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:53082835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGAAGTATAATGGAAGGCATGGA[C/T]GCAACATGCTGTGCCGTTTAGAAGA
- Phenotype
-
MIM: 611908
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RFT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74401054] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RFT1
- Gene Name
- RFT1 homolog
View Full Product Details