Product Details
- SNP ID
-
rs35529217
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97031432 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGACTGGAAACATGGGCTTTTA[A/G]AACACACACAGGGCTCTCCTTTAGC
- Phenotype
-
MIM: 603308
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTSV
PubMed Links
Gene Details
- Gene
- CTSV
- Gene Name
- cathepsin V
View Full Product Details