Product Details

SNP ID
rs10513467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:155032003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACATGCAGATAAACTAATGTAG[C/G]TATTTCCAAGGTACCCCATTTTCAC
Phenotype
MIM: 120520
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MME PubMed Links

Gene Details

Gene
MME
Gene Name
membrane metallo-endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000902.3 Intron NP_000893.2
NM_007287.2 Intron NP_009218.2
NM_007288.2 Intron NP_009219.2
NM_007289.2 Intron NP_009220.2
XM_006713646.3 Intron XP_006713709.1
XM_006713647.3 Intron XP_006713710.1
XM_011512855.2 Intron XP_011511157.1
XM_011512856.2 Intron XP_011511158.1
XM_011512857.2 Intron XP_011511159.1

View Full Product Details