Product Details

SNP ID
rs6596471
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:139369899 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAATATAGCAGTAGAATAACTCC[A/G]TTGCCCATCCTTGCTTCTTAATCAC
Phenotype
MIM: 605604 MIM: 603790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAIP2 PubMed Links

Gene Details

Gene
PAIP2
Gene Name
poly(A) binding protein interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033112.2 Intron NP_001028284.1
NM_016480.4 Intron NP_057564.3
XM_011543428.1 Intron XP_011541730.1
XM_017009522.1 Intron XP_016865011.1
XM_017009523.1 Intron XP_016865012.1
Gene
SLC23A1
Gene Name
solute carrier family 23 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005847.4 Intron NP_005838.3
NM_152685.3 Intron NP_689898.2
XM_005272148.3 Intron XP_005272205.3
XM_005272149.3 Intron XP_005272206.3
XM_006714741.2 Intron XP_006714804.2
XM_011543765.2 Intron XP_011542067.1
XM_011543766.1 Intron XP_011542068.1
XM_011543767.1 Intron XP_011542069.1
XM_011543768.1 Intron XP_011542070.1
XM_011543769.1 Intron XP_011542071.1

View Full Product Details