Product Details

SNP ID
rs719023
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:85591365 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACCTTAGGGCAGCTGTGCATTGA[C/T]TGGGTAGACGCCATTCTGGAGCAGG
Phenotype
MIM: 612490 MIM: 616757 MIM: 607029 MIM: 603177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF181 PubMed Links

Gene Details

Gene
RNF181
Gene Name
ring finger protein 181
There are no transcripts associated with this gene.

Gene
TMEM150A
Gene Name
transmembrane protein 150A
There are no transcripts associated with this gene.

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006634.2 Intron NP_006625.1
Gene
VAMP8
Gene Name
vesicle associated membrane protein 8
There are no transcripts associated with this gene.

View Full Product Details