Product Details

SNP ID
rs11693993
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:208360498 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GACCCGGCTGAGACTGTGCTTGTGC[A/G]GCGAGTGGAGGCCGCTCCCGGGGCC
Phenotype
MIM: 609414 MIM: 601469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIKFYVE PubMed Links

Gene Details

Gene
PIKFYVE
Gene Name
phosphoinositide kinase, FYVE-type zinc finger containing
There are no transcripts associated with this gene.

Gene
PTH2R
Gene Name
parathyroid hormone 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309516.1 Intron NP_001296445.1
NM_005048.3 Intron NP_005039.1

View Full Product Details