Product Details

SNP ID
rs36102758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53887666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCATGTGGTTTCCTTTATCTGG[A/G]ATAACTTCCTCTGATGTTCACCTCC
Phenotype
MIM: 607916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTR8 PubMed Links

Gene Details

Gene
ACTR8
Gene Name
ARP8 actin-related protein 8 homolog
There are no transcripts associated with this gene.

Gene
SELK
Gene Name
selenoprotein K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021237.3 Intron NP_067060.2

View Full Product Details