Product Details

SNP ID
rs2289385
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:8650366 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCAGGTTCTAGTTGGTACATTCAG[A/C]AATAACTAGGAGGGTTGTGCTAAAA
Phenotype
MIM: 601103
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFAP5 PubMed Links

Gene Details

Gene
MFAP5
Gene Name
microfibrillar associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297709.1 Intron NP_001284638.1
NM_001297710.1 Intron NP_001284639.1
NM_001297711.1 Intron NP_001284640.1
NM_001297712.1 Intron NP_001284641.1
NM_003480.3 Intron NP_003471.1

View Full Product Details