Product Details

SNP ID
rs2289384
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:8650278 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCATGACCCTTCTTCCTCCAGCGC[C/T]CACCTTCATTTCCCTTCTAAGTTCT
Phenotype
MIM: 601103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFAP5 PubMed Links

Gene Details

Gene
MFAP5
Gene Name
microfibrillar associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297709.1 Intron NP_001284638.1
NM_001297710.1 Intron NP_001284639.1
NM_001297711.1 Intron NP_001284640.1
NM_001297712.1 Intron NP_001284641.1
NM_003480.3 Intron NP_003471.1

View Full Product Details