Product Details

SNP ID
rs2255112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:54973664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAATTATTACCGTTGCTAAAGAA[G/T]CTTATTAACCACTACTATGTGTCAA
Phenotype
MIM: 604475
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EML6 PubMed Links

Gene Details

Gene
EML6
Gene Name
echinoderm microtubule associated protein like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039753.2 Intron NP_001034842.2
XM_017004098.1 Intron XP_016859587.1
XM_017004099.1 Intron XP_016859588.1
XM_017004100.1 Intron XP_016859589.1
XM_017004101.1 Intron XP_016859590.1
XM_017004102.1 Intron XP_016859591.1
Gene
RTN4
Gene Name
reticulon 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321859.1 Intron NP_001308788.1
NM_001321860.1 Intron NP_001308789.1
NM_001321861.1 Intron NP_001308790.1
NM_001321862.1 Intron NP_001308791.1
NM_001321863.1 Intron NP_001308792.1
NM_001321904.1 Intron NP_001308833.1
NM_007008.2 Intron NP_008939.1
NM_020532.4 Intron NP_065393.1
NM_153828.2 Intron NP_722550.1
NM_207520.1 Intron NP_997403.1
NM_207521.1 Intron NP_997404.1
XM_005264434.3 Intron XP_005264491.1
XM_017004518.1 Intron XP_016860007.1
XM_017004519.1 Intron XP_016860008.1

View Full Product Details