Product Details

SNP ID
rs2755253
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:67005160 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGTAGTCTGGAGGAGCTCACAGATC[C/T]GACCACTCCTGCCCTGTCCTCCCTG
Phenotype
MIM: 610804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35D1 PubMed Links

Gene Details

Gene
SLC35D1
Gene Name
solute carrier family 35 member D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015139.2 Intron NP_055954.1
XM_006710478.1 Intron XP_006710541.1
XM_011541070.1 Intron XP_011539372.1

View Full Product Details