Product Details

SNP ID
rs34442172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76377630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTCCAAGTCAGCGTTTCTCCA[G/T]TTCCACCGTGCTTTCATTTGTGTAC
Phenotype
MIM: 602167
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESRRB PubMed Links

Gene Details

Gene
ESRRB
Gene Name
estrogen related receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004452.3 Intron NP_004443.3
XM_011536547.2 Intron XP_011534849.1
XM_011536550.2 Intron XP_011534852.1
XM_011536553.2 Intron XP_011534855.1
XM_011536554.2 Intron XP_011534856.1
XM_017021085.1 Intron XP_016876574.1
XM_017021086.1 Intron XP_016876575.1

View Full Product Details