Product Details

SNP ID
rs11897415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223762702 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCAACATCTAGCCACCACACGCT[A/C]AGAATAAGCTTTGAGCATTTGGAGA
Phenotype
MIM: 615781
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP1S3 PubMed Links

Gene Details

Gene
AP1S3
Gene Name
adaptor related protein complex 1 sigma 3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039569.1 Intron NP_001034658.1
XM_011510600.2 Intron XP_011508902.1

View Full Product Details