Product Details

SNP ID
rs2167419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51752944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGCCATCTTAGGCCATGGGCT[A/G]GACAAGCTTAGCTTAGAACCAAGAC
Phenotype
MIM: 136537 MIM: 136538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FPR1 PubMed Links

Gene Details

Gene
FPR1
Gene Name
formyl peptide receptor 1
There are no transcripts associated with this gene.

Gene
FPR2
Gene Name
formyl peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005738.1 Intron NP_001005738.1
NM_001462.3 Intron NP_001453.1
XM_006723120.3 Intron XP_006723183.1
XM_017026542.1 Intron XP_016882031.1

View Full Product Details