Product Details

SNP ID
rs36007371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6832317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGAGGTTATCTATGGCTAGGAC[C/T]AGGGCATCAAGCAGGCGTTCTCAGT
Phenotype
MIM: 139130 MIM: 610342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNB3 PubMed Links

Gene Details

Gene
GNB3
Gene Name
G protein subunit beta 3
There are no transcripts associated with this gene.

Gene
GPR162
Gene Name
G protein-coupled receptor 162
There are no transcripts associated with this gene.

Gene
P3H3
Gene Name
prolyl 3-hydroxylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014262.4 Intron NP_055077.2

View Full Product Details