Product Details

SNP ID
rs2838678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44770927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGGGCCCACAGGGAACCAGGA[A/G]GGACCCAGAAAGGGTGCCGGGCCCT
Phenotype
MIM: 603124
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2G2 PubMed Links

Gene Details

Gene
UBE2G2
Gene Name
ubiquitin conjugating enzyme E2 G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202489.1 954 UTR 3 NP_001189418.1
NM_003343.5 954 UTR 3 NP_003334.2
NM_182688.2 954 UTR 3 NP_872630.1
XM_017028469.1 954 UTR 3 XP_016883958.1

View Full Product Details